RIE156331253 Rieke Metals, Inc.

4-(5,5-dimethyl-1,3-dioxan-2-yl)-4'-ethylbutyrophenone

  • MDL number MFCD03844277
  • Cas Number : 898787-21-8
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-3-3A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-3-3A
Availability details for
Quantity
  • Availability
  • Add to Cart