RIE156331254 Rieke Metals, Inc.

4-(5,5-Dimethyl-1,3-dioxan-2-yl)-4'-n-propylbutyrophenone

  • MDL number MFCD03844278
  • Cas Number : 898787-23-0
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-3-3B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-3-3B
Availability details for
Quantity
  • Availability
  • Add to Cart