RIE156331257 Rieke Metals, Inc.

4-(5,5-dimethyl-1,3-dioxan-2-yl)-4'-hexylbutyrophenone

  • MDL number MFCD03844281
  • Cas Number : 898787-29-6
  • Molecular Weight : 346.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-3-3E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-3-3E
Availability details for
Quantity
  • Availability
  • Add to Cart