RIE156331262 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-4'-pentylvalerophenone

  • MDL number MFCD03844286
  • Cas Number : 898755-48-1
  • Molecular Weight : 346.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-4-3D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-4-3D
Availability details for
Quantity
  • Availability
  • Add to Cart