RIE156331263 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-4'-hexylvalerophenone

  • MDL number MFCD03844287
  • Cas Number : 898755-51-6
  • Molecular Weight : 360.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-4-3E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-4-3E
Availability details for
Quantity
  • Availability
  • Add to Cart