RIE156331267 Rieke Metals, Inc.

4'-tert-butyl-4-(5,5-dimethyl-1,3-dioxan-2-yl)butyrophenone

  • MDL number MFCD03844292
  • Cas Number : 898755-67-4
  • Molecular Weight : 318.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8530-3-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8530-3-3
Availability details for
Quantity
  • Availability
  • Add to Cart