RIE156331275 Rieke Metals, Inc.

bis-3',5'-trifluoromethyl-4-(5,5-dimethyl-1,3-dioxan-2-yl)butyrophenone

  • MDL number MFCD03844306
  • Cas Number : 898755-91-4
  • Molecular Weight : 398.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8537-3-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8537-3-0
Availability details for
Quantity
  • Availability
  • Add to Cart