RIE156331278 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-5'-fluoro-2'-methylvalerophenone

  • MDL number MFCD03844309
  • Cas Number : 898755-98-1
  • Molecular Weight : 308.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8538-4-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8538-4-0
Availability details for
Quantity
  • Availability
  • Add to Cart