RIE156331291 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-1'-valeronaphthone

  • MDL number MFCD03844324
  • Cas Number : 898756-32-6
  • Molecular Weight : 326.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8546-4-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8546-4-0
Availability details for
Quantity
  • Availability
  • Add to Cart