RIE156331292 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-2'-propionaphthone

  • MDL number MFCD03844325
  • Cas Number : 898756-35-9
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8547-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8547-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart