RIE156331300 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-2'-phenylvalerophenone

  • MDL number MFCD03844332
  • Cas Number : 898756-59-7
  • Molecular Weight : 352.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8549-4-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8549-4-1
Availability details for
Quantity
  • Availability
  • Add to Cart