RIE156331301 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-4'-phenylpropiophenone

  • MDL number MFCD02261852
  • Cas Number : 898756-62-2
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8550-2-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8550-2-3
Availability details for
Quantity
  • Availability
  • Add to Cart