RIE156331305 Rieke Metals, Inc.

4'-Cyano-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261855
  • Cas Number : 898756-72-4
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8551-2-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8551-2-3
Availability details for
Quantity
  • Availability
  • Add to Cart