RIE156331309 Rieke Metals, Inc.

4'-cyano-5-(5,5-dimethyl-1,3-dioxan-2-yl)valerophenone

  • MDL number MFCD03844338
  • Cas Number : 898756-80-4
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8551-4-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8551-4-3
Availability details for
Quantity
  • Availability
  • Add to Cart