RIE156331313 Rieke Metals, Inc.

2',4'-dichloro-5-(5,5-dimethyl-1,3-dioxan-2-yl)valerophenone

  • MDL number MFCD03844342
  • Cas Number : 898756-90-6
  • Molecular Weight : 345.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8553-4-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8553-4-0
Availability details for
Quantity
  • Availability
  • Add to Cart