RIE156331319 Rieke Metals, Inc.

3',4'-dichloro-4-(5,5-dimethyl-1,3-dioxan-2-yl)butyrophenone

  • MDL number MFCD03844347
  • Cas Number : 898757-04-5
  • Molecular Weight : 331.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8556-3-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8556-3-0
Availability details for
Quantity
  • Availability
  • Add to Cart