RIE156331323 Rieke Metals, Inc.

2'-Chloro-3-(1,3-dioxan-2-yl)-4'-nitropropiophenone

  • MDL number MFCD02261868
  • Cas Number : 937040-35-2
  • Molecular Weight : 299.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8564-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8564-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart