RIE156331324 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-3'-iodo-4'-methylpropiophenone

  • MDL number MFCD02261873
  • Cas Number : 898757-13-6
  • Molecular Weight : 360.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8569-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8569-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart