RIE156331325 Rieke Metals, Inc.

4-(5,5-dimethyl-1,3-dioxan-2-yl)-3'-iodo-4'-methylbutyrophenone

  • MDL number MFCD03844363
  • Cas Number : 898757-15-8
  • Molecular Weight : 402.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8569-3-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8569-3-0
Availability details for
Quantity
  • Availability
  • Add to Cart