RIE156331333 Rieke Metals, Inc.

4'-chloro-2',5'-difluoro-3-(1,3-dioxan-2-yl)-propiophenone

  • MDL number MFCD02261883
  • Cas Number : 898757-38-5
  • Molecular Weight : 290.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8583-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8583-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart