RIE156331339 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-2'-[(4-trifluoromethyl)phenyl]propiophenone

  • MDL number MFCD03844378
  • Cas Number : 898757-56-7
  • Molecular Weight : 364.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8586-2-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8586-2-1
Availability details for
Quantity
  • Availability
  • Add to Cart