RIE156331347 Rieke Metals, Inc.

1-Chloro-3-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117648
  • Cas Number : 842123-93-7
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8035A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8035A
Availability details for
Quantity
  • Availability
  • Add to Cart