RIE156331350 Rieke Metals, Inc.

1-Chloro-3-fluoro-2-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117650
  • Cas Number : 842124-01-0
  • Molecular Weight : 216.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8037A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8037A
Availability details for
Quantity
  • Availability
  • Add to Cart