RIE156331357 Rieke Metals, Inc.

3-(1,3-Dioxan-2-yl)-5'-fluoro-2'-methylpropiophenone

  • MDL number MFCD02261840
  • Cas Number : 884504-39-6
  • Molecular Weight : 252.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8538-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8538-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart