RIE156331364 Rieke Metals, Inc.

3-(4-Methylphenyl)propionaldehyde

  • MDL number MFCD04112993
  • Cas Number : 5406-12-2
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8130

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8130
Availability details for
Quantity
  • Availability
  • Add to Cart