RIE156331365 Rieke Metals, Inc.

3-(3-Methylphenyl)propionaldehyde

  • MDL number MFCD07772907
  • Cas Number : 95416-60-7
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8131

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8131
Availability details for
Quantity
  • Availability
  • Add to Cart