RIE156331368 Rieke Metals, Inc.

3-(4-Chlorophenyl)propionaldehyde

  • MDL number MFCD07772912
  • Cas Number : 75677-02-0
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/8133

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8133
Availability details for
Quantity
  • Availability
  • Add to Cart