RIE156331372 Rieke Metals, Inc.

3',5'-dimethoxy-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261846
  • Cas Number : 884504-44-3
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8544-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8544-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart