RIE156331373 Rieke Metals, Inc.

Ethyl 4-(4-Methoxy-3-methylphenyl)-4-oxobutanoate

  • MDL number MFCD09801995
  • Cas Number : 2954-68-9
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7739B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7739B
Availability details for
Quantity
  • Availability
  • Add to Cart