RIE156331376 Rieke Metals, Inc.

4-(Ethylthio)benzaldehyde

  • MDL number MFCD01318145
  • Cas Number : 84211-94-9
  • Molecular Weight : 166.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8140

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8140
Availability details for
Quantity
  • Availability
  • Add to Cart