RIE156331379 Rieke Metals, Inc.

4-Phenylbutyraldehyde

  • MDL number MFCD00800383
  • Cas Number : 18328-11-5
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8143

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8143
Availability details for
Quantity
  • Availability
  • Add to Cart