RIE156331384 Rieke Metals, Inc.

1-bromo-2,4-dimethoxy-5-(1,3-dioxolan-2-yl)benzene

  • MDL number MFCD06209473
  • Cas Number : 552845-84-8
  • Molecular Weight : 289.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8505

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8505
Availability details for
Quantity
  • Availability
  • Add to Cart