RIE156331386 Rieke Metals, Inc.

1-Bromo-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD06209038
  • Cas Number : 52157-62-7
  • Molecular Weight : 235.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8507

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8507
Availability details for
Quantity
  • Availability
  • Add to Cart