RIE156331390 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261787
  • Cas Number : 167644-49-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8513-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8513-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart