RIE156331396 Rieke Metals, Inc.

2',3'-Dichloro-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261856
  • Cas Number : 884504-45-4
  • Molecular Weight : 289.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8552-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8552-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart