RIE156331403 Rieke Metals, Inc.

2-(1,3-Dioxolan-2-ylmethyl)-1-methoxybenzene

  • MDL number MFCD04117674
  • Cas Number : 842123-92-6
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8074A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8074A
Availability details for
Quantity
  • Availability
  • Add to Cart