RIE156331405 Rieke Metals, Inc.

3-(1,3-Dioxolan-2-ylmethyl)-1-methoxybenzene

  • MDL number MFCD04117675
  • Cas Number : 842123-95-9
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8075A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8075A
Availability details for
Quantity
  • Availability
  • Add to Cart