RIE156331415 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-4'-pentylpropiophenone

  • MDL number MFCD02261819
  • Cas Number : 884504-34-1
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-2-3D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-2-3D
Availability details for
Quantity
  • Availability
  • Add to Cart