RIE156331416 Rieke Metals, Inc.

4'-bromo-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261793
  • Cas Number : 376637-07-9
  • Molecular Weight : 299.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8515-2-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8515-2-3
Availability details for
Quantity
  • Availability
  • Add to Cart