RIE156331417 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-ylmethyl)-4-methylbenzene

  • MDL number MFCD01924578
  • Cas Number : 4412-50-4
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8083A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8083A
Availability details for
Quantity
  • Availability
  • Add to Cart