RIE156331422 Rieke Metals, Inc.

4-(2-Tetrahydro-2H-pyranoxy)benzaldehyde

  • MDL number MFCD02261777
  • Cas Number : 74189-56-3
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8092

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8092
Availability details for
Quantity
  • Availability
  • Add to Cart