RIE156331431 Rieke Metals, Inc.

1,3-Difluoro-4-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117656
  • Cas Number : 842124-12-3
  • Molecular Weight : 200.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8049A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8049A
Availability details for
Quantity
  • Availability
  • Add to Cart