RIE156331434 Rieke Metals, Inc.

1,2-Difluoro-4-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117658
  • Cas Number : 760211-54-9
  • Molecular Weight : 200.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8051A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8051A
Availability details for
Quantity
  • Availability
  • Add to Cart