RIE156331435 Rieke Metals, Inc.

1-Bromo-4-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117645
  • Cas Number : 4410-16-6
  • Molecular Weight : 243.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8031A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8031A
Availability details for
Quantity
  • Availability
  • Add to Cart