RIE156331438 Rieke Metals, Inc.

2-(1,3-Dioxan-2-yl)benzaldehyde

  • MDL number MFCD07700185
  • Cas Number : 139086-86-5
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8590

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8590
Availability details for
Quantity
  • Availability
  • Add to Cart