RIE183967908 Rieke Metals, Inc.

3,5-Dimethyl-4-methoxyphenethylzinc bromide

  • MDL number MFCD09965864
  • Molecular Weight : 308.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2266

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2266
Availability details for
Quantity
  • Availability
  • Add to Cart