RIE183967909 Rieke Metals, Inc.

4-Chloro-2-Methoxycarbonylphenylzinc bromide

  • MDL number MFCD09965865
  • Cas Number : 2056114-48-6
  • Molecular Weight : 314.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2267

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2267
Availability details for
Quantity
  • Availability
  • Add to Cart