RIE183967914 Rieke Metals, Inc.

4-Fluoro-3-methylbenzylzinc chloride

  • MDL number MFCD11226463
  • Molecular Weight : 223.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2272

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2272
Availability details for
Quantity
  • Availability
  • Add to Cart