RIE183967916 Rieke Metals, Inc.

4-Chloro-2-methylbenzylzinc chloride

  • MDL number MFCD11226465
  • Molecular Weight : 240.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2274

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2274
Availability details for
Quantity
  • Availability
  • Add to Cart