RIE183967919 Rieke Metals, Inc.

5-Fluoro-2-methoxybenzylzinc chloride

  • MDL number MFCD11226468
  • Molecular Weight : 239.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2277

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2277
Availability details for
Quantity
  • Availability
  • Add to Cart